Specialty vials
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5180-0841 5180-0844 5182-3454 5183-2030 5183-2073 5183-4497 5183-4510 5184-3550 5184-3551 5184-3552 5184-3554 5184-3555 5184-3556
HEWL5180-0841EA 238 EUR
HEWL5180-0841 HEWL5180-0844 HEWL5182-3454 HEWL5183-2030 HEWL5183-2073 HEWL5183-4497 HEWL5183-4510 HEWL5184-3550 HEWL5184-3551 HEWL5184-3552 HEWL5184-3554 HEWL5184-3555 HEWL5184-3556
Specialty vials
flesjes
These speciality vials with wide opening are made of type I borosilicate glass.
High recovery glass vials are used for sample concentration and injection without transferring to microvolume inserts.
Micro-V vials are wineglass shaped used for microsampling.