Washers, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F728-16 F728-08
BOHLF728-16EA 5.1 EUR
BOHLF728-16 BOHLF728-08
Washers, BOLA
Afdichtingen Afdichtringen
PA
  • Temperature resistance: from –40 to +100 °C
For stabilising the flange and avoiding damages when tightening the tube end fitting.
Order Now
Specificaties
Environmentally Preferable