PIG® HAM-O® universal mat pads and rolls
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT116 MAT267 MAT268 MAT269
NEWPMAT116EA 355 EUR
NEWPMAT116 NEWPMAT267 NEWPMAT268 NEWPMAT269
PIG® HAM-O® universal mat pads and rolls
Absorptiematerialen en bescherming tegen morsen
"Hamouflage" pattern hides grime so mat always looks clean. Scuff-resistant top layer stands up to foot traffic in aisles and walkways.
  • Exclusive "hamouflage" top layer saves on product and disposal costs; absorbed grime is invisible, so you'll leave mat in place longer and save money
  • Fast-wicking "diaper action" transfers liquids from the top layer into the absorbent inner layers; spills disappear in seconds
  • Tough, spunbond top layer stands up to heavy foot traffic; won't rip, tear, fray or create a trip hazard, even when saturated
  • Under the Ham-O top layer is the proven eight-layer, 100% polypropylene construction
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Absorbs and retains oils, coolants, solvents and water
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
They can be wrung out and incinerated after use to reduce waste or for fuels blending. The heavy-weight construction delivers greater durability and absorbency for demanding tasks and large-volume cleanups. Rolls are ideal for walkways and large areas to soak up leaks and overspray.