PIG® spill bag
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KITE251 KITE351 KITE451
121-0066EA 30.4 EUR
121-0066 121-0070 121-0074
PIG® spill bag
Absorptiematerialen en bescherming tegen morsen
High-visibility, compact spill bag travels anywhere you do for spill response on the go.
  • Brightly coloured yellow bag features a reflective stripe for greater visibility and easy identification
  • Drawstring closure is ideal for carrying or hanging on a wall
  • Packed with a variety of PIG® absorbents to handle small spills
PIG® spill bag is flexible and small enough to fit behind the seat of a lorry; stows almost anywhere and is easy to carry in the event of a spill
Opgelet: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.