PIG® Extra-Duty universal mat pads and rolls
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT173 MAT175
NEWPMAT173EA 131 EUR
NEWPMAT173 NEWPMAT175
PIG® Extra-Duty universal mat pads and rolls
Absorptiematerialen en bescherming tegen morsen
Both sides of this absorbent mat are tough enough for rough surfaces. PIG Extra-Duty Mat is highly abrasion resistant so it won't rip, tear or fray.
  • Special spunbond outer layers resist ripping and fraying from wiping rough surfaces or parts that would shred conventional mats
  • Eight layers of 100% polypropylene are thermally bonded; won't rip, tear or fray even when saturated
  • Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
  • Absorbs and retains oils, coolants, solvents and water
  • Low-lint for wiping rough parts or