PIG® STAT-MAT® absorbent mat pads and rolls
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT214 MAT215 MAT402 MAT414
NEWPMAT214EA 157 EUR
NEWPMAT214 NEWPMAT215 NEWPMAT402 NEWPMAT414
PIG® STAT-MAT® absorbent mat pads and rolls
Absorptiematerialen en bescherming tegen morsen
Specially treated to reduce the risk of fueling explosions sparked by static.
  • STAT-MAT Pads and Rolls reduce the risk of electrostatic discharge; a critical precaution around flammable liquids and vapours
  • Perfect for fueling areas and solvent cleanup
  • Eight layers of 100% polypropylene are thermally bonded; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
  • Packaged in special static-