Spill containment barriers, SPILLBLOCKER® Dike
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PLR253-NG PLR254-NG PLR267-YW PLR268-BK
NEWPPLR253-NGEA 504 EUR
NEWPPLR253-NG NEWPPLR254-NG NEWPPLR267-YW NEWPPLR268-BK
Spill containment barriers, SPILLBLOCKER® Dike
Absorptiematerialen en bescherming tegen morsen
Flexible portable 100% polyurethane barrier for containment of spillages on floors.
  • Barrier contains or diverts the flow of liquids, without absorbing the liquid
  • Self-contained dove tail connectors allow for multiple lengths to contain larger spills
  • Range of profile heights
These reusable barriers can be easily cleaned with soap and water.