PIG® sump skimmers
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SKM403 SKM404
NEWPSKM403EA 240 EUR
NEWPSKM403 NEWPSKM404
PIG® sump skimmers
Absorptiematerialen en bescherming tegen morsen
Keep sumps, tanks and bilges oil-free with easy-to-retrieve PIG Sump Skimmers.
  • Simply lower this floating skimmer into any water-based liquid to absorb oils without taking in water
  • Ideal for soaking up unwanted oil-based fluids that collect in sumps, tanks and bilges
  • Polypropylene loop can be attached to rope (not included) for easy deployment and retrieval
  • Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water
  • Bright white colour makes absorbed oil easier to see; clearly shows saturation level
Can be incinerated after use to reduce waste or for fuels blending. Polypropylene skin resists chemicals and tearing; meets NFPA 99 standards for static decay.