illustra™ Ready-To-Go™ GenomiPhi™ V3 DNA amplification kits
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25-6601-24 25-6601-96 25-6601-97
25-6601-24EA 238 EUR
25-6601-24 25-6601-96 25-6601-97
illustra™ Ready-To-Go™ GenomiPhi™ V3 DNA amplification kits
Reagentia voor nucleïnezuren Isothermal Amplification Reagents
illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 12 to 20 μg yield from nanogram amounts of DNA sample.
  • Representative amplification of the whole genome
  • Preformulated, predispensed, single dose, lyophilised cakes for rapid reaction set-up and greater reproducibility
  • Quick and simple automation friendly protocol; no thermal cycler required
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint
  • Yields high quality DNA (eg. for high-throughput genotyping and DNA archival)
Leveringsinformatie: illustra Ready-To-Go GenomiPhi V3 DNA Amplification Kit contains all of the components necessary for whole genome amplification by isothermal strand displacement amplification. The reaction mixture is provided as predispensed, single-dose, lyophilised cakes in either strips of 8 tubes, or 96-well plates.
Order Now
Specificaties
Environmentally Preferable
Applicatie