illustra™ Ready-To-Go™ GenomiPhi™ HY DNA amplification kits
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25-6603-24 25-6603-96
25-6603-24EA 568 EUR
25-6603-24 25-6603-96
illustra™ Ready-To-Go™ GenomiPhi™ HY DNA amplification kits
Reagentia voor nucleïnezuren Isothermal Amplification Reagents
illustra™ Ready-To-Go™ GenomiPhi™ HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
  • Representative amplification of the whole genome
  • Preformulated, predispensed, single dose, lyophilised cakes for rapid reaction set-up and greater reproducibility
  • Quick and simple automation-friendly protocol; no thermal cycler required
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint
  • Yields high quality DNA (eg. for high-throughput genotyping and DNA archival)