Tetrabutylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Tetrabutylammoniumtetrafluoroborate
T0914-25G
T0914-500G
T0914-100G
TCIAT0914-25GEA
55
EUR
TCIAT0914-25G
TCIAT0914-500G
TCIAT0914-100G
Tetrabutylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Tetrabutylammonium tetrafluoroborate
Formule:
C₁₆H₃₆BF₄N MW: 329.27 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011634 CAS nummer: 429-42-5 EINECS: 207-058-8 UN: 1759 ADR: 8 III |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 160.0 to 163.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...