Cesium bromide ≥99.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Cesiumbromide
C2202-25G C2202-100G
TCIAC2202-25GEA 55 EUR
TCIAC2202-25G TCIAC2202-100G
Cesium bromide ≥99.0% (by titrimetric analysis)
Cesium bromide
Formule: BrCs
MW: 212.81 g/mol
Kookpunt: 1300
Smeltpunt: 636°C
Dichtheid: 4.46
MDL Nummer: MFCD00010954
CAS nummer: 7787-69-1
EINECS: 232-130-0
Merck Index: 14,02009
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d56dea47e1fb407a8c8a57525f35648e10e9dab&vid=5d56dea47e1fb407a8c8a57525f35648e10e9dab&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White powder to crystal
Purity min. 99.0 %