2-Bromo-4-methylpyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Bromo-4-picoline
B2112-25G B2112-5G
TCIAB2112-25GEA 94.6 EUR
TCIAB2112-25G TCIAB2112-5G
2-Bromo-4-methylpyridine ≥98.0% (by GC)
2-Bromo-4-methylpyridine
Formule: C₆H₆BrN
Kookpunt: 42 - 44 °C (0,6 -rr)
Smeltpunt: 218…228 °C
Dichtheid: 1,545 g/cm³ (20 °C)
MDL Nummer: MFCD00082590
CAS nummer: 4926-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d599944501be0af10873922aeceb9fcb06ab6a6&vid=3d599944501be0af10873922aeceb9fcb06ab6a6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %