1-Bromo-3,5-dimethyladamantane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B1821-25G
B1821-5G
TCIAB1821-25GEA
125
EUR
TCIAB1821-25G
TCIAB1821-5G
1-Bromo-3,5-dimethyladamantane ≥98.0% (by GC)
1-Bromo-3,5-dimethyladamantane
Formule:
C₁₂H₁₉Br Kookpunt: 273°C Vlampunt: 109°C |
MDL Nummer:
MFCD00077197 CAS nummer: 941-37-7 EINECS: 213-378-9 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...