2-Bromo-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3243-25G B3243-5G
TCIAB3243-25GEA 153 EUR
TCIAB3243-25G TCIAB3243-5G
2-Bromo-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
2-Bromo-N,N-dimethylaniline
Formule: C₈H₁₀BrN
Kookpunt: 108°C
Smeltpunt: 38 °C
Dichtheid: 1,388 g/cm³ (25 °C)
MDL Nummer: MFCD00013522
CAS nummer: 698-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b99c392edf83bb5b3b32b25c9021e2cc4dec8e0&vid=3b99c392edf83bb5b3b32b25c9021e2cc4dec8e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR