2-Bromophenyl isocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
I0498-25G I0498-5G
TCIAI0498-25GEA 350 EUR
TCIAI0498-25G TCIAI0498-5G
2-Bromophenyl isocyanate ≥98.0% (by GC)
2-Bromophenyl isocyanate
Formule: C₇H₄BrNO
Kookpunt: 62 - 64 °C (0,5 -rr)
Dichtheid: 1,6 g/cm³ (20 °C)
Vlampunt: 107°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00001995
CAS nummer: 1592-00-3
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfeeefe9ccbf53e80b3c0c2911d3f6ea56f168a7&vid=bfeeefe9ccbf53e80b3c0c2911d3f6ea56f168a7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %