2-Amino-3,5-dibromopyridine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 3,5-Dibromo-2-pyridylamine, 3,5-Dibromopyridin-2-amine
A1588-25G A1588-5G
TCIAA1588-25GEA 81.4 EUR
TCIAA1588-25G TCIAA1588-5G
2-Amino-3,5-dibromopyridine ≥98.0% (by GC)
2-Amino-3,5-dibromopyridine
Formule: C₅H₄Br₂N₂
Kookpunt: 254 °C (1013 hPa)
Smeltpunt: 104 °C
MDL Nummer: MFCD00038041
CAS nummer: 35486-42-1
EINECS: 252-590-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a2d8cd56b14134778d12618641d161916c67c46&vid=9a2d8cd56b14134778d12618641d161916c67c46&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C