2-Amino-3-bromo-5-methylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Bromo-5-methylpyridin-2-amine, 2-Amino-3-bromo-5-picoline, 3-Bromo-5-methyl-2-pyridylamine

A1555-25G A1555-5G
TCIAA1555-25GEA 134 EUR
TCIAA1555-25G TCIAA1555-5G
2-Amino-3-bromo-5-methylpyridine ≥98.0% (by GC)
2-Amino-3-bromo-5-methylpyridine
Formule: C₆H₇BrN₂
MDL Nummer: MFCD00068231
CAS nummer: 17282-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4822372a38ce868bb8eb66172697ba78e8411adf&vid=4822372a38ce868bb8eb66172697ba78e8411adf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 72.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR