1-Bromo-2-methoxyethane ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Bromoethyl methyl ether
B1242-25G B1242-250G
TCIAB1242-25GEA 37.4 EUR
TCIAB1242-25G TCIAB1242-250G
1-Bromo-2-methoxyethane ≥95.0% (by GC)
1-Bromo-2-methoxyethane
Formule: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Kookpunt: 114°C
Vlampunt: 28°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000236
CAS nummer: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed7d1608900732e25427fece9e7bf942f450ca5e&vid=ed7d1608900732e25427fece9e7bf942f450ca5e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %