2-Bromothioanisole ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2023-25G B2023-5G
TCIAB2023-25GEA 151 EUR
TCIAB2023-25G TCIAB2023-5G
2-Bromothioanisole ≥96.0% (by GC)
2-Bromothioanisole
Formule: C₇H₇BrS
Kookpunt: 256°C
Dichtheid: 1,5135 g/cm³ (16 °C)
MDL Nummer: MFCD00000066
CAS nummer: 19614-16-5
EINECS: 243-183-4
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=596dfd4d85f3fae2c06bd54224efb28933d1d586&vid=596dfd4d85f3fae2c06bd54224efb28933d1d586&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Brown clear liquid
Purity (GC) min. 96.0 %