4-(Methylthio)benzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M0739-25G M0739-250G
TCIAM0739-25GEA 31.9 EUR
TCIAM0739-25G TCIAM0739-250G
4-(Methylthio)benzaldehyde ≥97.0% (by GC)
4-(Methylthio)benzaldehyde
Formule: C₈H₈OS
Kookpunt: 153°C
Dichtheid: 1,14 g/cm³ (20 °C)
MDL Nummer: MFCD00006948
CAS nummer: 3446-89-7
EINECS: 222-365-7
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=438e248d685cd12a91147234463f29ceda039496&vid=438e248d685cd12a91147234463f29ceda039496&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %