1,4-Dichloro-2-butyne ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D1528-25G D1528-5G D1528-100G
TCIAD1528-25GEA 128 EUR
TCIAD1528-25G TCIAD1528-5G TCIAD1528-100G
1,4-Dichloro-2-butyne ≥97.0% (by GC)
1,4-Dichloro-2-butyne
Formule: C₄H₄Cl₂
Kookpunt: 166°C
Dichtheid: 1,2613 g/cm³ (20 °C)
Vlampunt: 160°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000981
CAS nummer: 821-10-3
EINECS: 212-474-8
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b8c273c7e2e3ae03b8a39be507d45efda4fd40a&vid=6b8c273c7e2e3ae03b8a39be507d45efda4fd40a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %