Trichloro(phenyl)silane ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
P0240-25G P0240-500G
TCIAP0240-25GEA 17.6 EUR
TCIAP0240-25G TCIAP0240-500G
Trichloro(phenyl)silane ≥98.0% (by GC)
Trichloro(phenyl)silane
Formule: C₆H₅Cl₃Si
Kookpunt: 201°C
Smeltpunt: –40 °C
Dichtheid: 1,315 g/cm³ (20 °C)
Vlampunt: 91°C
MDL Nummer: MFCD00000479
CAS nummer: 98-13-5
EINECS: 202-640-8
UN: 1804
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18fd8e484900fb37f1da954108c3a44c3b391cf3&vid=18fd8e484900fb37f1da954108c3a44c3b391cf3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %