2,4,5-Trichloropyrimidine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T2512-25G T2512-5G
TCIAT2512-25GEA 236 EUR
TCIAT2512-25G TCIAT2512-5G
2,4,5-Trichloropyrimidine ≥98.0% (by GC)
2,4,5-Trichloropyrimidine
Formule: C₄HCl₃N₂
Kookpunt: 96°C
MDL Nummer: MFCD03788200
CAS nummer: 5750-76-5
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cde982627eed476c51e647903dd629de19ac469d&vid=cde982627eed476c51e647903dd629de19ac469d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR