2,4-Dichloro-5-methylpyrimidine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D3613-1G D3613-25G D3613-5G
TCIAD3613-1GEA 33 EUR
TCIAD3613-1G TCIAD3613-25G TCIAD3613-5G
2,4-Dichloro-5-methylpyrimidine ≥98.0% (by GC)
2,4-Dichloro-5-methylpyrimidine
Formule: C₅H₄Cl₂N₂
Kookpunt: 109°C
Smeltpunt: 26 °C
Dichtheid: 1,39 g/cm³ (25 °C)
MDL Nummer: MFCD00023197
CAS nummer: 1780-31-0
EINECS: 217-227-8
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17b1833fe7179da6c04de6b7736474f0eeb3eba6&vid=17b1833fe7179da6c04de6b7736474f0eeb3eba6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %