5-Chloro-1-indanone ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C1644-1G C1644-5G
TCIAC1644-1GEA 37.4 EUR
TCIAC1644-1G TCIAC1644-5G
5-Chloro-1-indanone ≥97.0% (by GC)
5-Chloro-1-indanone
Formule: C₉H₇ClO
MW: 166.61 g/mol
Kookpunt: 150
Smeltpunt: 100
MDL Nummer: MFCD00041456
CAS nummer: 42348-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae657af745026833a612cfd3208d9915c9635a63&vid=ae657af745026833a612cfd3208d9915c9635a63&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 93.0 to 97.0 deg-C