Dichlon ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D0384-25G
D0384-500G
TCIAD0384-25GEA
17.6
EUR
TCIAD0384-25G
TCIAD0384-500G
Dichlon ≥95.0% (by GC)
Dichlon
Formule:
C₁₀H₄Cl₂O₂ Kookpunt: 304 °C (1013 hPa) Smeltpunt: 193 °C Dichtheid: 1,44 g/cm³ (16 °C) |
MDL Nummer:
MFCD00001677 CAS nummer: 117-80-6 EINECS: 204-210-5 UN: 2811 ADR: 6.1,III Merck Index: 14,03045 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 194.0 to 198.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...