2-Chloro-5-fluorobenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C2080-1G C2080-5G
TCIAC2080-1GEA 31.9 EUR
TCIAC2080-1G TCIAC2080-5G
2-Chloro-5-fluorobenzonitrile ≥98.0% (by GC)
2-Chloro-5-fluorobenzonitrile
Formule: C₇H₃ClFN
MDL Nummer: MFCD03094168
CAS nummer: 57381-56-3
EINECS: 260-714-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b74ee43c8c01c573f20a833cd6df4eaeecf4bbd4&vid=b74ee43c8c01c573f20a833cd6df4eaeecf4bbd4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C