n-Butyronitril ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B0768-25ML
B0768-500ML
TCIAB0768-25MLEA
17.6
EUR
TCIAB0768-25ML
TCIAB0768-500ML
n-Butyronitril ≥99.0% (by GC)
n-Butyronitril
Formule:
C₄H₇N Kookpunt: 118 °C (1013 hPa) Smeltpunt: –112 °C Dichtheid: 0,79 g/cm³ (20 °C) Vlampunt: 17°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001968 CAS nummer: 109-74-0 EINECS: 203-700-6 UN: 2411 ADR: 3,II Merck Index: 14,01597 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...