Benzyl cyanide ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
P0128-25ML
P0128-500ML
TCIAP0128-25MLEA
19.8
EUR
TCIAP0128-25ML
TCIAP0128-500ML
Benzyl cyanide ≥98.0% (by GC)
Benzyl cyanide
Formule:
(C₆H₅)CH₂CN Kookpunt: 230°C Smeltpunt: –24°C Dichtheid: 1,02 g/cm³ (20 °C) Vlampunt: 101°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001894 CAS nummer: 140-29-4 EINECS: 205-410-5 UN: 2470 ADR: 6.1,III Merck Index: 14,01131 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...