Tricyclo[5.2.1.0(2,6)]-8-decanone ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
T1321-25ML T1321-500ML
TCIAT1321-25MLEA 29.7 EUR
TCIAT1321-25ML TCIAT1321-500ML
Tricyclo[5.2.1.0(2,6)]-8-decanone ≥95.0% (by GC)
Tricyclo[5.2.1.0(2,6)]-8-decanone
Formule: C₁₀H₁₄O
Kookpunt: 132°C
Vlampunt: 102°C
MDL Nummer: MFCD00168034
CAS nummer: 13380-94-4
EINECS: 236-458-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69dbec0cf515de01f6fef807b731a08e082b082&vid=d69dbec0cf515de01f6fef807b731a08e082b082&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %