Chalcone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1,3-Diphenyl-2-propen-1-one, Phenyl Styryl ketone, 2-Benzalacetophenone, 2-Benzylideneacetophenone

C0071-25G C0071-500G C0071-100G
TCIAC0071-25GEA 24.2 EUR
TCIAC0071-25G TCIAC0071-500G TCIAC0071-100G
Chalcone ≥98.0% (by GC)
Chalcone
Formule: (C₆H₅)C(O)CHCH(C₆H₅)
Kookpunt: 345 °C (1013 hPa)
Smeltpunt: 53…56 °C
Dichtheid: 1,0712 g/cm³ (62.4 °C)
MDL Nummer: MFCD00003082
CAS nummer: 94-41-7
EINECS: 202-330-2
Merck Index: 14,02037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cbead705aba34d5394bfa33302b32af94fe1d06&vid=9cbead705aba34d5394bfa33302b32af94fe1d06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR