O-Acetylsalicylic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®
A2262-25G A2262-500G
TCIAA2262-25GEA 17.6 EUR
TCIAA2262-25G TCIAA2262-500G
O-Acetylsalicylic acid ≥98.0% (by GC, titration analysis)
O-Acetylsalicylic acid
Formule: 2-(CH₃CO₂)C₆H₄CO₂H
MW: 180.16 g/mol
Kookpunt: 140
Vlampunt: 250
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002430
CAS nummer: 50-78-2
EINECS: 200-064-1
UN: 2811
ADR: 6.1 III
Merck Index: 14,00851
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cc8faa34585dc16f5db6704f890dece94b219b6&vid=0cc8faa34585dc16f5db6704f890dece94b219b6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 133.0 to 138.0 deg-C
Specificaties
Environmentally Preferable