2-Ethyl-9,10-anthraquinone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-Ethylanthracene-9,10-dione, 2-Ethylanthraquinone
E0063-25G E0063-500G
TCIAE0063-25GEA 19.8 EUR
TCIAE0063-25G TCIAE0063-500G
2-Ethyl-9,10-anthraquinone ≥98.0% (by GC)
2-Ethyl-9,10-anthraquinone
Formule: C₁₆H₁₂O₂
MW: 236.27 g/mol
Kookpunt: 415
MDL Nummer: MFCD00001237
CAS nummer: 84-51-5
EINECS: 201-535-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba3697594347a5d3c81fbc851e5f121982ca1e2&vid=aba3697594347a5d3c81fbc851e5f121982ca1e2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C
Solubility in hot Methanol almost transparency