Benzoyl bromide ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
B0104-25G
B0104-5G
TCIAB0104-25GEA
160
EUR
TCIAB0104-25G
TCIAB0104-5G
Benzoyl bromide ≥98.0% (by GC, titration analysis)
Benzoyl bromide
Formule:
C₇H₅BrO Kookpunt: 219°C Smeltpunt: 0 °C Dichtheid: 1,527 to 1.5564 g/cm³ (10 °C) Vlampunt: 90°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000112 CAS nummer: 618-32-6 EINECS: 210-544-2 UN: 3265 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...