4-Biphenylcarbonyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
P1079-25G P1079-5G
TCIAP1079-25GEA 138 EUR
TCIAP1079-25G TCIAP1079-5G
4-Biphenylcarbonyl chloride ≥98.0% (by GC, titration analysis)
4-Biphenylcarbonyl chloride
Formule: C₁₃H₉ClO
Kookpunt: 160°C
Smeltpunt: 110…112 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000692
CAS nummer: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=874d4967bc236d23f4b7886cd9df77055fb967eb&vid=874d4967bc236d23f4b7886cd9df77055fb967eb&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C