Fumaroyl chloride ≥95.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
F0152-25G F0152-500G
TCIAF0152-25GEA 44 EUR
TCIAF0152-25G TCIAF0152-500G
Fumaroyl chloride ≥95.0% (by GC, titration analysis)
Fumaroyl chloride
Formule: C₄H₂Cl₂O₂
Kookpunt: 161 °C (1013 hPa)
Smeltpunt: –2 °C
Dichtheid: 1,41 g/cm³ (20 °C)
Vlampunt: 77°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000733
CAS nummer: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5052ce0a769231068e6255ba8deded48fc9bb3&vid=da5052ce0a769231068e6255ba8deded48fc9bb3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %