Diallylftalaat ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Phthalic acid diallyl ester

P0290-25ML P0290-500ML
TCIAP0290-25MLEA 18.7 EUR
TCIAP0290-25ML TCIAP0290-500ML
Diallylftalaat ≥98.0% (by GC)
Diallylftalaat
Formule: C₁₄H₁₄O₄
Kookpunt: 158°C
Smeltpunt: –70 °C
Dichtheid: 1,12 g/cm³ (20 °C)
Vlampunt: 163°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008646
CAS nummer: 131-17-9
EINECS: 205-016-3
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3efd66b55dece19785e821a6f0fd0089d3ac1137&vid=3efd66b55dece19785e821a6f0fd0089d3ac1137&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR