Ethyl phenylacetate ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P0122-25ML P0122-500ML
TCIAP0122-25MLEA 17.6 EUR
TCIAP0122-25ML TCIAP0122-500ML
Ethyl phenylacetate ≥99.0% (by GC)
Ethyl phenylacetate
Formule: C₁₀H₁₂O₂
Kookpunt: 226°C
Smeltpunt: –29 °C
Dichtheid: 1,03 g/cm³ (20 °C)
Vlampunt: 107°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009178
CAS nummer: 101-97-3
EINECS: 202-993-8
Merck Index: 14,03840
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a62e1c2fc6ea24f42e6d2d2f28e36bb49059bbc&vid=9a62e1c2fc6ea24f42e6d2d2f28e36bb49059bbc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %