Dibutyl maleate ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Maleic acid dibutyl ester
M0009-25G
M0009-500G
TCIAM0009-25GEA
17.6
EUR
TCIAM0009-25G
TCIAM0009-500G
Dibutyl maleate ≥95.0% (by GC)
Dibutyl maleate
Formule:
C₁₂H₂₀O₄ MW: 228.29 g/mol Kookpunt: 161°C Smeltpunt: –85°C Vlampunt: 140°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009447 CAS nummer: 105-76-0 EINECS: 203-328-4 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |