1-Naphthylacetic acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Planofix, 1-Naphthaleneacetic acid
N0005-25G N0005-500G
TCIAN0005-25GEA 17.6 EUR
TCIAN0005-25G TCIAN0005-500G
1-Naphthylacetic acid ≥98.0% (by titrimetric analysis)
1-Naphthylacetic acid
Formule: C₁₂H₁₀O₂
MW: 186.21 g/mol
Kookpunt: 352
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004046
CAS nummer: 86-87-3
EINECS: 201-705-8
Merck Index: 14,06371
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a19ce715825eddc954d91493a8ac53e48236b24&vid=2a19ce715825eddc954d91493a8ac53e48236b24&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 129.0 to 135.0 deg-C