Sebacic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Decanedioic acid, 1,8-Octanedicarboxylic acid, 1,10-Decanedioic acid
S0022-25G
S0022-500G
TCIAS0022-25GEA
18.7
EUR
TCIAS0022-25G
TCIAS0022-500G
Sebacic acid ≥98.0% (by GC, titration analysis)
Sebacic acid
Formule:
C₁₀H₁₈O₄ MW: 202.25 g/mol Kookpunt: 374 Vlampunt: 221 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004440 CAS nummer: 111-20-6 EINECS: 203-845-5 Bereik: 01-2119519212-52 Merck Index: 14,08415 |
|
Specification Test Results
Appearance | White powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...