Phenyl salicylate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
S0017-25G S0017-500G
TCIAS0017-25GEA 17.6 EUR
TCIAS0017-25G TCIAS0017-500G
Phenyl salicylate ≥98.0% (by GC)
Phenyl salicylate
Formule: C₁₃H₁₀O₃
Kookpunt: 173°C
Smeltpunt: 41…43 °C
Dichtheid: 1,307 g/cm³ (20 °C)
Vlampunt: 160 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002213
CAS nummer: 118-55-8
EINECS: 204-259-2
ADR: 9,III
Merck Index: 14,07310
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d190f2c627160ab825dd3186ff7725875aa0ee48&vid=d190f2c627160ab825dd3186ff7725875aa0ee48&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 44.0 deg-C
Solubility in Methanol almost transparency