Difluoroacetic acid ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D1423-25G D1423-5G
TCIAD1423-25GEA 132 EUR
TCIAD1423-25G TCIAD1423-5G
Difluoroacetic acid ≥97.0% (by titrimetric analysis)
Difluoroacetic acid
Formule: C₂H₂F₂O₂
Kookpunt: 135°C
Smeltpunt: 0°C
Dichtheid: 1,526 g/cm³ (20 °C)
Vlampunt: 78°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004220
CAS nummer: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7cc635bc44758434b6df660a2c472cad4ae80d7&vid=d7cc635bc44758434b6df660a2c472cad4ae80d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Yellow to Orange clear liquid
Purity (Neutralization titration) min. 97.0 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR