2-Fluoro-3-iodopyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0733-1G F0733-5G
TCIAF0733-1GEA 30.8 EUR
TCIAF0733-1G TCIAF0733-5G
2-Fluoro-3-iodopyridine ≥98.0% (by GC)
2-Fluoro-3-iodopyridine
Formule: C₅H₃FIN
Kookpunt: 99°C
MDL Nummer: MFCD03095287
CAS nummer: 113975-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17f36c58d22d8f568e81336746094f2fb9703e97&vid=17f36c58d22d8f568e81336746094f2fb9703e97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR