N,N,N'-Triethylethylenediamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0743-25ML T0743-5ML
TCIAT0743-25MLEA 146 EUR
TCIAT0743-25ML TCIAT0743-5ML
N,N,N'-Triethylethylenediamine ≥98.0% (by GC, titration analysis)
N,N,N'-Triethylethylenediamine
Formule: C₈H₂₀N₂
Kookpunt: 145°C
Dichtheid: 0,804 g/cm³ (20 °C)
Vlampunt: 32°C
MDL Nummer: MFCD00009054
CAS nummer: 105-04-4
EINECS: 203-264-7
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf8316341b429a73b8f6d894a18306f6ae4af99d&vid=cf8316341b429a73b8f6d894a18306f6ae4af99d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR