Phenyl isothiocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
I0191-25G I0191-500G I0191-100G
TCIAI0191-25GEA 24.2 EUR
TCIAI0191-25G TCIAI0191-500G TCIAI0191-100G
Phenyl isothiocyanate ≥98.0% (by GC)
Phenyl isothiocyanate
Formule: C₇H₅NS
Kookpunt: 71°C
Smeltpunt: –21 °C
Dichtheid: 1,13 g/cm³ (20 °C)
Vlampunt: 88°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004798
CAS nummer: 103-72-0
EINECS: 203-138-1
UN: 2922
ADR: 8,II
Merck Index: 14,07297
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d9dfdf55d3b2bb4203c62229d1fbfebb054a7d0&vid=7d9dfdf55d3b2bb4203c62229d1fbfebb054a7d0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specificaties
Environmentally Preferable