1-Aminopyrene ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0632-1G
A0632-25G
A0632-5G
TCIAA0632-1GEA
50.6
EUR
TCIAA0632-1G
TCIAA0632-25G
TCIAA0632-5G
1-Aminopyrene ≥98.0% (by HPLC, titration analysis)
1-Aminopyrene
Formule:
C₁₆H₁₁N Kookpunt: 415 °C (1013 hPa) Smeltpunt: 118…119 °C |
MDL Nummer:
MFCD00004140 CAS nummer: 1606-67-3 EINECS: 216-521-3 |
|
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...