2-Aminothiophenol ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0267-25ML A0267-500ML A0267-100ML
TCIAA0267-25MLEA 20.9 EUR
TCIAA0267-25ML TCIAA0267-500ML TCIAA0267-100ML
2-Aminothiophenol ≥97.0% (by titrimetric analysis)
2-Aminothiophenol
Formule: C₆H₇NS
Kookpunt: 234°C
Smeltpunt: 26 °C
Dichtheid: 1,17 g/cm³ (20 °C)
Vlampunt: 122°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007702
CAS nummer: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=277a6cc6050051bbd6a56bf6942baf2fdeb0dc42&vid=277a6cc6050051bbd6a56bf6942baf2fdeb0dc42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR