6-Benzylaminopurine ≥99.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1088-25G
B1088-5G
TCIAB1088-25GEA
50.6
EUR
TCIAB1088-25G
TCIAB1088-5G
6-Benzylaminopurine ≥99.0% (by HPLC, titration analysis)
6-Benzylaminopurine
Formule:
C₁₂H₁₁N₅ Smeltpunt: 230…234 °C |
MDL Nummer:
MFCD00005572 CAS nummer: 1214-39-7 EINECS: 214-927-5 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in 0.1mol/L NaOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...